L(-)-Tyrosine

Supplier: PanReac AppliChem

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

A3401.1000
A3401.1000EA 465 EUR
A3401.1000
L(-)-Tyrosine
L(-)-Tyrosine
Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR