Spectinomycin dihydrochloride pentahydrate ≥95% (by titrimetric analysis), BioChemica®

Supplier: PanReac AppliChem

Synonyms: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

A3834.0005 A3834.0010
A3834.0005EA 151 EUR
A3834.0005 A3834.0010
Spectinomycin dihydrochloride pentahydrate ≥95% (by titrimetric analysis), BioChemica®
Spectinomycin dihydrochloride pentahydrate
Formula: C₁₄H₂₄N₂O₇·2HCl·5H₂O
MW: 495.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150886
CAS Number: 22189-32-8
EINECS: 244-554-3

Order Now

Specification Test Results

Assay (Titrimetric) Min. 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR