Cell Meter™ Fluorimetric Intracellular Peroxynitrite Assay Kit *Optimized for Flow Cytometry*
Supplier: AAT BIOQUEST
16317
AATB16317EA 537 EUR
AATB16317
Cell Meter™ Fluorimetric Intracellular Peroxynitrite Assay Kit *Optimized for Flow Cytometry*
Assays Cellular Assays
Peroxynitrite (ONOO-) is a strong oxidizing species and a highly active nitrating agent.
  • High sensitivity
  • Bright fluorescence
  • Suitable for live cell imaging
Caution: For research use only.