MagaDye™ 561 ddATP
Supplier: AAT BIOQUEST
17062 17066
AATB17062EA 1410 EUR
AATB17062 AATB17066
MagaDye™ 561 ddATP
Nucleic Acid Reagents Sanger Sequencing Reagents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more