ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Supplier: AAT BIOQUEST
17207
AATB17207EA 292 EUR
AATB17207
ddCTP [2',3'-Dideoxycytidine-5'-triphosphate] *10 mM in ddH₂O* (1 µmole)
Nucleic Acid Reagents Sanger Sequencing Reagents
Sanger sequencing, also known as the chain termination method, is a technique for DNA sequencing based upon the selective incorporation of chain-terminating dideoxynucleotides (ddNTPs) by DNA polymerase.
  • In-house quality control
  • Useful for sanger sequencing applications
Caution: For research use only.