PUR-1
Supplier: AAT BIOQUEST
17560
AATB17560EA 152 EUR
AATB17560
PUR-1
Assays Cellular Assays
PUR-1 is essentially non-fluorescent in the absence of nucleic acids.
  • Useful For Multicolor Imaging
  • Bright Fluorescence
  • 40% Fluorescence Quantum Yield
Caution: For research use only.