Portelite™ Fluorimetric DNA Quantitation Kit with Broad Dynamic Range *Optimized for Cytocite™ and Qubit™ Fluorometers*
Supplier: AAT BIOQUEST
17665
AATB17665EA 120 EUR
AATB17665
Portelite™ Fluorimetric DNA Quantitation Kit with Broad Dynamic Range *Optimized for Cytocite™ and Qubit™ Fluorometers*
Assays Nucleic Acid Assays
DNA Quantitation is a very important task in DNA sample preparations for various analyses.
  • Linear over three orders of magnitude
  • Exceptional dsDNA selectivity over rna
  • Robust protocol
  • Multiple applications
Caution: For research use only.