Anti-WAPAL Rabbit Monoclonal Antibody [clone: EPR3407]

Supplier: Abcam
AB109537-100
ABCAAB109537-100EA 848 EUR
ABCAAB109537-100
Anti-WAPAL Rabbit Monoclonal Antibody [clone: EPR3407]
Antibodies
Type: Primary
Antigen: WAPAL (wings apart-like homolog (Drosophila))
Clonality: monoclonal
Clone: EPR3407
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR