Anti-CRLF2 Rabbit polyclonal antibody unconjugated

Supplier: Abcam
AB109626-50
ABCAAB109626-50EA 939 EUR
ABCAAB109626-50
Anti-CRLF2 Rabbit polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CRLF2 (cytokine receptor-like factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR