Anti-PRDX1 Goat Polyclonal Antibody

Supplier: Abcam
AB109766-50
ABCAAB109766-50EA 931 EUR
ABCAAB109766-50
Anti-PRDX1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PRDX1 (peroxiredoxin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR