Anti-KRT18 Rabbit Monoclonal Antibody [clone: SP69]

Supplier: Abcam
AB93741-100
ABCAAB93741-100EA 947 EUR
ABCAAB93741-100
Anti-KRT18 Rabbit Monoclonal Antibody [clone: SP69]
Antibodies
Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of

More Product Information
Order Now

SPECIFICATIONS

Antigen symbol KRT18
Antigen name keratin 18
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone SP69
Reactivity
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR