Anti-POU2F1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP11702C
ABGEAP11702CEA 354 EUR
ABGEAP11702C
Anti-POU2F1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: POU2F1 (POU class 2 homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR