Anti-CPSF7 Rabbit Polyclonal Antibody

Supplier: Abgent
AP12151B
ABGEAP12151BEA 354 EUR
ABGEAP12151B
Anti-CPSF7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CPSF7 (cleavage and polyadenylation specific factor 7, 59kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CPSF7
Environmentally Preferable
Antigen name V-Ets Avian Erythroblastosis Virus E2 Oncogene Homolog 1
Antigen synonyms EWSR2|ETS-1|p54|c-ets-1
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype
Western blot Yes
ELISA Yes
Epitope C-terminal
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR