Anti-XIRP1 Rabbit Polyclonal Antibody

Supplier: Abgent
AP13612B
ABGEAP13612BEA 354 EUR
ABGEAP13612B
Anti-XIRP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: XIRP1 (xin actin-binding repeat containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR