Anti-SULT1C2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP2602B
ABGEAP2602BEA 354 EUR
ABGEAP2602B
Anti-SULT1C2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SULT1C2 (sulfotransferase family, cytosolic, 1C, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR