Anti-TACSTD2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP6681C
ABGEAP6681CEA 354 EUR
ABGEAP6681C
Anti-TACSTD2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TACSTD2 (tumor-associated calcium signal transducer 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR