Anti-NAB2 Rabbit Polyclonal Antibody

Supplier: Abgent
AP7635C
ABGEAP7635CEA 354 EUR
ABGEAP7635C
Anti-NAB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NAB2 (NGFI-A binding protein 2 (EGR1 binding protein 2))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR