Anti-INSR Rabbit Polyclonal Antibody

Supplier: Abgent
AP7654B
ABGEAP7654BEA 354 EUR
ABGEAP7654B
Anti-INSR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: INSR (insulin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR