Anti-UBE3A Mouse Monoclonal Antibody [clone: 2F6]

Supplier: Abnova
H00007337-M01
ABNOH00007337-M01EA 397 EUR
ABNOH00007337-M01
Anti-UBE3A Mouse Monoclonal Antibody [clone: 2F6]
Antibodies
Type: Primary
Antigen: UBE3A
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR