Anti-SLC17A7 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S28-9]

Supplier: Abnova
MAB11528
ABNOMAB11528EA 912 EUR
ABNOMAB11528
Anti-SLC17A7 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S28-9]
Antibodies
Mouse monoclonal antibody raised against recombinant of Slc17a7.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC17A7
Clonality: Monoclonal
Clone: S28-9
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR