Anti-CD4 Mouse Monoclonal Antibody [clone: HCD4] (FITC)

Supplier: Abnova
MAB23586.500UG
ABNOMAB23586.500UGEA 423 EUR
ABNOMAB23586.500UG
Anti-CD4 Mouse Monoclonal Antibody [clone: HCD4] (FITC)
Antibodies
Order Now

SPECIFICATIONS

Antigen symbol CD4
Environmentally Preferable
Clonality Monoclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Clone HCD4
Reactivity Human
Host Mouse
Isotype
Immunogen Mouse FITC monoclonal antibody raised against synthetic peptide of human CD4.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR