Anti-Mycoplasma bovis Mouse Monoclonal Antibody [clone: MY11-320.1]

Supplier: Abnova
MAB8330
ABNOMAB8330EA 402 EUR
ABNOMAB8330
Anti-Mycoplasma bovis Mouse Monoclonal Antibody [clone: MY11-320.1]
Antibodies
Mouse monoclonal antibody raised against Mycoplasma bovis.

Type: Primary
Antigen: Mycoplasma bovis
Clonality: Monoclonal
Clone: MY11-320.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR