Anti-Hepatitis A virus VP1 Mouse Monoclonal Antibody [clone: 54d64]

Supplier: Abnova
MAB8950
ABNOMAB8950EA 523 EUR
ABNOMAB8950
Anti-Hepatitis A virus VP1 Mouse Monoclonal Antibody [clone: 54d64]
Antibodies
Mouse monoclonal antibody raised against partial recombinant Hepatitis A virus VP1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: 54d64
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR