Anti-SFRP4 Goat Polyclonal Antibody

Supplier: Abnova
PAB15663
ABNOPAB15663EA 523 EUR
ABNOPAB15663
Anti-SFRP4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of SFRP4.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 4-6 ?g/ml

Type: Primary
Antigen: SFRP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR