Anti-Rb Rabbit Recombinant Antibody [clone: R09-8G7]

Supplier: Abnova
RAB01678.100UL
ABNORAB01678.100ULEA 648 EUR
ABNORAB01678.100UL
Anti-Rb Rabbit Recombinant Antibody [clone: R09-8G7]
Antibodies
Order Now

SPECIFICATIONS

Antigen symbol Rb
Environmentally Preferable
Clonality Recombinant
Clone R09-8G7
Reactivity Mouse
Host Rabbit
Isotype
Immunogen Recombinant rabbit monoclonal antibody raised against mouse Rb

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR