Avantor® ACE® C18-AR, HPLC Columns, 3 µm
Supplier: Avantor
ACE-119-0102GDEA 263 EUR
ACE-119-0102GD ACE-119-0103GD ACE-119-0202 ACE-119-0203 ACE-119-0302 ACE-119-0303 ACE-119-0501 ACE-119-0502 ACE-119-0503 ACE-119-0504 ACE-119-0546 ACE-119-1002 ACE-119-1003 ACE-119-1004 ACE-119-1046 ACE-119-1203 ACE-119-1204 ACE-119-1502 ACE-119-1503 ACE-119-1504 ACE-119-1546 ACE-119-7502 ACE-119-7503 ACE-119-7546 ACE-119-05003 ACE1191546MVK
Avantor® ACE® C18-AR, HPLC Columns, 3 µm
Chromatography Columns
The Avantor® ACE® C18-AR phase utilises a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and