4-tert-Butylcyclohexanol mixture of isomers 99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hexahydro-p-tert-butylphenol, 4-(tert-Butyl)cyclohexanol

107900050 107902500
ACRO107900050EA 19.7 EUR
ACRO107900050 ACRO107902500
4-tert-Butylcyclohexanol mixture of isomers 99%
4-tert-Butylcyclohexanol
Formula: C₁₀H₂₀O
Boiling Pt: 224 °C
Melting Pt: 60…69 °C
Density: 0,87 g/cm³ (80 °C)
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00001473
CAS Number: 98-52-2
EINECS: 202-676-4

Order Now

Specification Test Results

Appearance White Powder or granules
Infrared spectrum Conforms
Melting point 60 to 69 °C (depending on cis/trans ratio)
GC ≥98.5 %
Ratio Cis/trans ratio: approximately 70% trans, 30% cis
Specific optical rotation -12° to -18° (20 °C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR