4-Chlorobenzyl alcohol 99%

Supplier: Thermo Fisher Scientific

Synonyms: (4-Chlorophenyl)methanol

108770250 108771000
ACRO108770250EA 47 EUR
ACRO108770250 ACRO108771000
4-Chlorobenzyl alcohol 99%
4-Chlorobenzyl alcohol
Formula: C₇H₇ClO
Boiling Pt: 234 °C (760 mmHg)
Melting Pt: 69…72 °C
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00004652
CAS Number: 873-76-7
EINECS: 212-852-2

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Melting point 69 to 72 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR