(±)-2,3-Epoxy-1-propanol 96%

Supplier: Thermo Fisher Scientific
Danger

120051000 120055000 120050025
ACRO120051000EA 52.6 EUR
ACRO120051000 ACRO120055000 ACRO120050025
(±)-2,3-Epoxy-1-propanol 96%
(±)-2,3-Epoxy-1-propanol
Formula: C₃H₆O₂
Boiling Pt: 160…161 °C (760 mmHg)
Melting Pt: –54 °C
Density: 1.11 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005147
CAS Number: 556-52-5
EINECS: 209-128-3
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4360 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR