Hydrochloric acid standard solution 1 mol/l (1 N) in water

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

124210010 124210025 124210100
ACRO124210010EA 52.7 EUR
ACRO124210010 ACRO124210025 ACRO124210100
Hydrochloric acid standard solution 1 mol/l (1 N) in water
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.02 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.998 to 1.002 N (20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR