Malonyl dichloride 97%

Supplier: Thermo Fisher Scientific
Danger

125280250 125281000
ACRO125280250EA 83.5 EUR
ACRO125280250 ACRO125281000
Malonyl dichloride 97%
Malonyl dichloride
Formula: C₃H₂Cl₂O₂
Boiling Pt: 47 °C (15 mmHg)
Density: 1.45 g/cm³
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000735
CAS Number: 1663-67-8
EINECS: 216-772-9
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4620 to 1.4660 (20°C, 589 nm)
Titration Argentometric ≥96.0 %
Residue after evaporation ≤0.0003 %
Conductivity ≤1 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR