N-Methylformanilide 99%

Supplier: Thermo Fisher Scientific
Warning

126810010 126811000 126815000 126810100
ACRO126810010EA 145 EUR
ACRO126810010 ACRO126811000 ACRO126815000 ACRO126810100
N-Methylformanilide 99%
N-Methylformanilide
Formula: C₈H₉NO
Boiling Pt: 243…244 °C (760 mmHg)
Melting Pt: 8…13 °C
Density: 1.09 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00003283
CAS Number: 93-61-8
EINECS: 202-262-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5600 to 1.5620 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR