Purine 99%

Supplier: Thermo Fisher Scientific

Synonyms: 1H-Purine

131670010 AC13167.1 AC13167.10 131670050
ACRO131670010EA 197 EUR
ACRO131670010 MAYBAC13167.1 MAYBAC13167.10 ACRO131670050
Purine 99%
Purine
Formula: C₅H₄N₄
Boiling Pt: 265 °C (1013 hPa)
Density: 1,22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079221
CAS Number: 120-73-0
EINECS: 204-421-2
Merck Index: 12,08129

Order Now

Specification Test Results

Appearance White to light yellow to beige Crystalline powder and/or chunks
Infrared spectrum Conforms
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR