Pyridine-2,5-dicarboxylic acid 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Isocinchomeronic acid, 2,5-Pyridinedicarboxylic acid

131870050
ACRO131870050EA 28.2 EUR
ACRO131870050
Pyridine-2,5-dicarboxylic acid 98%
Pyridine-2,5-dicarboxylic acid
Formula: C₇H₅NO₄
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006297
CAS Number: 100-26-5
EINECS: 202-834-2
Merck Index: 13,05177

Order Now

Specification Test Results

Appearance Yellow to green Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.1 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR