Resorcinol 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,3-Dihydroxybenzene, 1,3-Benzenediol

132292500 132290500 132290010 132290050
ACRO132292500EA 79.7 EUR
ACRO132292500 ACRO132290500 ACRO132290010 ACRO132290050
Resorcinol 98%
Resorcinol
Formula: C₆H₆O₂
Boiling Pt: 281 °C (760 mmHg)
Melting Pt: 109…111 °C
Density: 1,272 g/cm³ (20 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00002269
CAS Number: 108-46-3
EINECS: 203-585-2
UN: 2876
ADR: 6.1,III
Merck Index: 12,08323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69fa2900537bdb42ccfd55e4318288eb881a61a0&vid=e62c617319ff4711a7ff00e69774ad92cfd75d09&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow to beige to pink Crystals or granules or flakes
Infrared spectrum Conforms
Melting point 108 to 112 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR