Spermidine 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

132740010 132740050 132740250 132741000
ACRO132740010EA 41.9 EUR
ACRO132740010 ACRO132740050 ACRO132740250 ACRO132741000
Spermidine 99%
Spermidine
Formula: C₇H₁₉N₃
Boiling Pt: 128…130 °C (14 mmHg)
Melting Pt: 22…25 °C
Density: 0.925 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008229
CAS Number: 124-20-9
EINECS: 204-689-0
UN: 3259
ADR: 8,II
Merck Index: 12,08893

Order Now

Specification Test Results

Appearance Clear colorless Liquid after melting
Infrared spectrum Conforms
Refractive index 1.4785 to 1.4805 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR