Chloroacetyl chloride 98%

Supplier: Thermo Fisher Scientific
Danger

147290010 147291000 147292500 147290025
ACRO147290010EA 90.3 EUR
ACRO147290010 ACRO147291000 ACRO147292500 ACRO147290025
Chloroacetyl chloride 98%
Chloroacetyl chloride
Formula: C₂H₂Cl₂O
Boiling Pt: 105 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.42 g/cm³
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000725
CAS Number: 79-04-9
EINECS: 201-171-6
UN: 1752
ADR: 6.1,
Merck Index: 13,02074

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR