Formic acid ≥98% 98+%, pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Aminic Acid, Methanoic acid

147930010 147930025 147930250 147932500 147930100
ACRO147930010EA 48.6 EUR
ACRO147930010 ACRO147930025 ACRO147930250 ACRO147932500 ACRO147930100
Formic acid ≥98% 98+%, pure
Formic acid
Formula: HCO₂H
Boiling Pt: 101 °C (760 mmHg)
Melting Pt: 8 °C
Density: 1.22 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3700 to 1.3720 (20°C, 589 nm)
Color scale ≤20 APHA
Titration with NaOH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR