Dipropyl ether 99+%

Supplier: Thermo Fisher Scientific
Danger

150040500
ACRO150040500EA 120 EUR
ACRO150040500
Dipropyl ether 99+%
Dipropyl ether
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 89
Melting Pt: –123 °C
Density: 0.73 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00009376
CAS Number: 111-43-3
EINECS: 203-869-6
UN: 2384
ADR: 3 II
Merck Index: 12,08042

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3790 to 1.3810 (20°C, 589 nm)
GC ≥99.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR