4-Chlorobenzaldehyde 98%

Supplier: Thermo Fisher Scientific
Warning

150440010 150441000 150442500
ACRO150440010EA 98 EUR
ACRO150440010 ACRO150441000 ACRO150442500
4-Chlorobenzaldehyde 98%
4-Chlorobenzaldehyde
Formula: C₇H₅ClO
Boiling Pt: 214 °C (760 mmHg)
Melting Pt: >46 °C
Density: 1.196 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003379
CAS Number: 104-88-1
EINECS: 203-247-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow Crystals or crystalline or fused solid
Infrared spectrum Conforms
Melting point 45 to 49 °C
GC ≥97.5 %
Free acid ≤1.5 % (as 4-Chlorobenzoic acid)
Elemental Analysis 17.45 to 18.90 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR