Bromocresol green sodium salt, water soluble indicator

Supplier: Thermo Fisher Scientific
Warning

151320050 151320250
ACRO151320050EA 30.2 EUR
ACRO151320050 ACRO151320250
Bromocresol green sodium salt, water soluble indicator
Bromocresol green sodium salt
Formula: C₂₁H₁₃Br₄NaO₅S
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148898
CAS Number: 67763-24-0
EINECS: 263-657-4

Order Now

Specification Test Results

Appearance Dark brown to green-black Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Visual transition interval from pH 3.8 (yellow) to pH 5.4 (blue)
Clarity of solution (0.1% in water) Clear


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR