1,2-Propylenediamine 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1,2-Diaminopropane

152400010 152400025 152402500
ACRO152400010EA 137 EUR
ACRO152400010 ACRO152400025 ACRO152402500
1,2-Propylenediamine 99%
1,2-Propylenediamine
Formula: C₃H₁₀N₂
Boiling Pt: 119…120 °C (760 mmHg)
Melting Pt: –37 °C
Density: 0.87 g/cm³
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00008089
CAS Number: 78-90-0
EINECS: 201-155-9
UN: 2258
ADR: 8,II
Merck Index: 12,08037

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %
Color scale ≤20 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR