Bromoethane 98%, pure

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Bromic ether, Hydrobromic ether

154210010 154215000 154210025
ACRO154210010EA 78.2 EUR
ACRO154210010 ACRO154215000 ACRO154210025
Bromoethane 98%, pure
Bromoethane
Formula: BrH₂CCH₃
Boiling Pt: 38.4 °C (1013 hPa)
Melting Pt: –119 °C
Density: 1.46 g/cm³
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000232
CAS Number: 74-96-4
EINECS: 200-825-8
UN: 1891
ADR: 6.1,II
Merck Index: 13,03807

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
Water ≤0.03 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR