(Methyl)triphenylphosphonium bromide 98%

Supplier: Thermo Fisher Scientific
Danger

156950100 156951000 156950025 156955000
ACRO156950100EA 25.3 EUR
ACRO156950100 ACRO156951000 ACRO156950025 ACRO156955000
(Methyl)triphenylphosphonium bromide 98%
(Methyl)triphenylphosphonium bromide
Formula: C₁₉H₁₈BrP
Melting Pt: 230…234 °C
Flash Pt: >240 °C
Storage Temperature: Ambient
MDL Number: MFCD00011804
CAS Number: 1779-49-3
EINECS: 217-218-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 230 to 234 °C
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR