2-Chloropyrazine 98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Chloropyrazine

158060100 158060500 AC15806.10 AC15806.50
ACRO158060100EA 60.9 EUR
ACRO158060100 ACRO158060500 MAYBAC15806.10 MAYBAC15806.50
2-Chloropyrazine 98%
2-Chloropyrazine
Formula: C₄H₃ClN₂
Boiling Pt: 153 °C (1013 hPa)
Density: 1.28 g/cm³
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00006124
CAS Number: 14508-49-7
EINECS: 238-517-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5390 (20°C, 589 nm)
Water ≤3 % (K.F.)
GC ≥97.5 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0005 meq/g
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR