D(+)-Tryptophan 99%

Supplier: Thermo Fisher Scientific

Synonyms: H-D-Trp-OH

168630050 168630250 168631000
ACRO168630050EA 44 EUR
ACRO168630050 ACRO168630250 ACRO168631000
D(+)-Tryptophan 99%
D(+)-Tryptophan
Formula: C₁₁H₁₂N₂O₂
Melting Pt: 282…285 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005647
CAS Number: 153-94-6
EINECS: 205-819-9
Merck Index: 14,09797

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
Solubility (1 % in water) Clear colorless to light yellow
Specific optical rotation +29.5° to +33.5° (20°C, 589 nm) (c=1, H2O)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR