Sodium metabisulphite, anhydrous 97%, extra pure

Supplier: Thermo Fisher Scientific
Danger

171490025 171490100
ACRO171490025EA 55.6 EUR
ACRO171490025 ACRO171490100
Sodium metabisulphite, anhydrous 97%, extra pure
Sodium metabisulphite
Formula: Na₂S₂O₅
Melting Pt: 150 °C (dec.)
Density: 1,48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167602
CAS Number: 7681-57-4
EINECS: 231-673-0
REACH: 01-2119531326-45
Merck Index: 12,08784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=110c9b08e7e977eb8c8a9a97bee16470ca9699c5&vid=004a574d6ed7e27d7cc82336f2179f23453618d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Titration Iodimetric ≥96.0 %
Thiosulphate (S2O3) ≤0.05 %
Iron (Fe) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR