Heptafluorobutyric acid 99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid

172800250 172801000 172805000
ACRO172800250EA 64.3 EUR
ACRO172800250 ACRO172801000 ACRO172805000
Heptafluorobutyric acid 99%
Heptafluorobutyric acid
Formula: C₄HF₇O₂
Boiling Pt: 120 °C (755 mmHg)
Melting Pt: –17.5 °C
Density: 1.64 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004171
CAS Number: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR