4,4'-Biphenol 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: [1,1'-Biphenyl]-4,4'-diol, 4,4'-Dihydroxybiphenyl, Biphenyl-4,4'-diol

173670250 173671000 173670100 173672500
ACRO173670250EA 89 EUR
ACRO173670250 ACRO173671000 ACRO173670100 ACRO173672500
4,4'-Biphenol 97%
4,4'-Biphenol
Formula: C₁₂H₁₀O₂
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 278…283 °C
Density: 1,25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002348
CAS Number: 92-88-6
EINECS: 202-200-5

Order Now

Specification Test Results

Appearance Off-white to light beige Powder
Infrared spectrum Conforms
Melting point 278 to 283 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR