2,5-Dimethylpyrazine 99%

Supplier: Thermo Fisher Scientific
Warning

174520050 174520250
ACRO174520050EA 50 EUR
ACRO174520050 ACRO174520250
2,5-Dimethylpyrazine 99%
2,5-Dimethylpyrazine
Formula: C₆H₈N₂
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: 15 °C
Density: 0.99 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00006147
CAS Number: 123-32-0
EINECS: 204-618-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4987 to 1.5007 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR